EMAIL THIS PAGE TO A FRIEND
Environmental monitoring and assessment
The use of rapidly synergistic cloud point extraction for the separation and preconcentration of trace amounts of Ni (II) ions from food and water samples coupling with flame atomic absorption spectrometry determination.
PMID 26857255
Abstract
A novel improved preconcentration method known as rapidly synergistic cloud point extraction (RS-CPE) was established for nickel preconcentration and determination prior to its determination by flame atomic absorption spectrometry. In this work, the traditional CPE pattern was changed and greatly simplified in order to be applicable in metal extraction and detection. This method was accomplished in room temperature in 1 min. Non-ionic surfactant Triton X-114 was used as extractant. Octanol worked as cloud point revulsant and synergic reagent. The various parameters affecting the extraction and preconcentration of nickel such as sample pH, 2,2'-Furildioxime concentration, amounts of octanol, amounts of Triton X-114, type of diluting solvent, extraction time, and ionic strength were investigated and optimized. Under optimal conditions, the calibration curve showed an excellent linearity in the concentration range of 2-200 μg L(-1), and the limit of detection was 0.6 μg L(-1) for nickel. The developed method was successfully applied for the determination of nickel in food and water samples. The results showed that, the proposed method can be used as a cheap, rapid, and efficient method for the extraction and preconcentration of nickel from real samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X114
Triton X-114, laboratory grade