EMAIL THIS PAGE TO A FRIEND
Food chemistry
Sphingoid esters from the molecular distillation of squid oil: A preliminary bioactivity determination.
PMID 26868543
Abstract
A mixture of sphingoid esters was isolated (1.4% w/w) from the molecular distillation of crude squid visceral oil. A preliminary investigation on the bioactivity profile and toxic potential of this residue was carried out by in vitro experiments. No cytotoxicity and a moderate lipase inhibition activity were highlighted.