EMAIL THIS PAGE TO A FRIEND
Oncology letters
Myeloid-derived suppressor cells enhance the expression of melanoma-associated antigen A4 in a Lewis lung cancer murine model.
PMID 26870289
Abstract
The cancer-testis (CT) family of antigens are expressed in multiple types of malignant neoplasm and are silent in normal tissues, apart from the testis. Immunotherapy targeting CT antigens is a promising therapeutic strategy for treatment of solid tumors. One member of this family, melanoma-associated antigen A4 (MAGE-A4), has been demonstrated to be expressed in melanomas and lung cancer. Patients with tumors expressing the MAGE-A4 antigen exhibit specific cellular and humoral immune responses to the antigen, resulting in a favorable prognosis. Conversely, the expression of MAGE-A4 is associated with poor survival in lung cancer. Myeloid-derived suppressor cells (MDSCs) are a heterogeneous population of immunosuppressive cells, which are upregulated in the cancer microenvironment. Little is known regarding any potential correlation between the expression of MAGE-A4 antigens and the accumulation of MDSCs. The present study aimed to examine the association between circulating MDSC levels and MAGE-A4 expression in a mouse model of Lewis lung cancer. The expression of MAGE-A4 in tumor cells or tissues was evaluated using western blotting, while the percentage of MDSCs (CD11b
Related Materials