EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
Heparanase Is Essential for the Development of Acute Experimental Glomerulonephritis.
PMID 26873445
Abstract
Heparanase, a heparan sulfate (HS)--specific endoglucuronidase, mediates the onset of proteinuria and renal damage during experimental diabetic nephropathy. Glomerular heparanase expression is increased in most proteinuric diseases. Herein, we evaluated the role of heparanase in two models of experimental glomerulonephritis, being anti-glomerular basement membrane and lipopolysaccharide-induced glomerulonephritis, in wild-type and heparanase-deficient mice. Induction of experimental glomerulonephritis led to an increased heparanase expression in wild-type mice, which was associated with a decreased glomerular expression of a highly sulfated HS domain, and albuminuria. Albuminuria was reduced in the heparanase-deficient mice in both models of experimental glomerulonephritis, which was accompanied by a better renal function and less renal damage. Notably, glomerular HS expression was preserved in the heparanase-deficient mice. Glomerular leukocyte and macrophage influx was reduced in the heparanase-deficient mice, which was accompanied by a reduced expression of both types 1 and 2 helper T-cell cytokines. In vitro, tumor necrosis factor-α and lipopolysaccharide directly induced heparanase expression and increased transendothelial albumin passage. Our study shows that heparanase contributes to proteinuria and renal damage in experimental glomerulonephritis by decreasing glomerular HS expression, enhancing renal leukocyte and macrophage influx, and affecting the local cytokine milieu.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU041011 MISSION® esiRNA, esiRNA human HPSE (esiRNA1)