EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Simultaneous removal of chromate and nitrate in a packed-bed bioreactor using biodegradable meal box as carbon source and biofilm carriers.
PMID 26896715
Abstract
An up-flow packed-bed bioreactor was constructed to investigate the simultaneous removal of chromate and nitrate using biodegradable meal box as carbon source and biofilm carriers. The bioreactor was operated for 164days with varying influent Cr(VI) concentrations (2.0-50.0mg/L) and hydraulic retention times (HRT, 10-24h). It was shown that complete denitrification and Cr(VI) reduction could be achieved when influent Cr(VI) concentrations were lower than 20mg/L with a HRT of 17h. Shortening the HRT could significantly reduce the effluent CODcr. It was also observed that Cr(III) was mainly immobilized on the biofilm. Further investigation on Cr distribution in the biofilm compartments indicated that Cr(VI) reduction occurred in all compartments and the intercellular Cr was dominant. High-throughput sequencing analysis showed that Proteobacteria, Bacteroidetes and Firmicutes were the dominant phyla in the biofilm and Cr(VI) stress had a negative effect on the abundance of most bacteria.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
80805
1,4-Di(maleimido)butane, ≥99.0% (HPLC)
C12H12N2O4