EMAIL THIS PAGE TO A FRIEND
Biomedicine and Pharmacotherapy
Pioglitazone up-regulates long non-coding RNA MEG3 to protect endothelial progenitor cells via increasing HDAC7 expression in metabolic syndrome.
PMID 26898430