EMAIL THIS PAGE TO A FRIEND
Archives of toxicology
Sterigmatocystin-induced checkpoint adaptation depends on Chk1 in immortalized human gastric epithelial cells in vitro.
PMID 26914363
Abstract
Sterigmatocystin (ST) is a common contaminant detected in food and animal feed that has been recognized as a possible human carcinogen. Our previous studies demonstrate that ST causes DNA damage and subsequently triggers cell cycle arrest in G