Journal of minimal access surgery
Ambulatory laparoscopic cholecystectomy: A single center experience.
PMID 26917919
To evaluate the demographic and clinical parameters affecting the outcomes of ambulatory laparoscopic cholecystectomy (ALC) in terms of pain, nausea, anxiety level, and satisfaction of patients in a tertiary health center. ALC was offered to 60 patients who met the inclusion criteria. Follow-up (questioning for postoperative pain or discomfort, nausea or vomiting, overall satisfaction) was done by telephone contact on the same day at 22:00 p.m. and the first day after surgery at 8: 00 a.m. and by clinical examination one week after operation. STAI I and II data were used for proceeding to the level of anxiety of patients before and/or after the operation. Sixty consecutive patients, with a mean age of 40.6 ± 8.1 years underwent ALC. Fifty-five (92%) patients could be sent to their homes on the same day but five patients could not be sent due to anxiety, pain, or social indications. Nausea was reported in four (6.7%) cases and not associated with any demographic or clinical features of patients. On the other hand, pain has been reported in 28 (46.7%) cases, and obesity and shorter duration of gallbladder disease were associated with the increased pain perception (P = 0.009 and 0.004, respectively). Preopereative anxiety level was significantly higher among patients who could not complete the ALC procedure (P = 0.018). Correct management of these possible adverse effects results in the increased satisfaction of patients and may encourage this more cost-effective and safe method of laparoscopic cholecystectomy.
Related Materials
Product #
Molecular Formula
Add to Cart
Remifentanil hydrochloride, ≥97% (HPLC), powder