EMAIL THIS PAGE TO A FRIEND
Food chemistry
Structural confirmation of novel oligosaccharides isolated from sugar beet molasses.
PMID 26920296
Abstract
Eleven oligosaccharides were isolated from sugar beet molasses using carbon-Celite column chromatography and HPLC. The constituent sugars and linkage positions were determined using methylation analysis, MALDI-TOF-MS, and NMR measurements. The configurations of isolated oligosaccharides were confirmed based on detailed NMR analysis. Based on our results, three of the 11 oligosaccharides were novel.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72555
1-Kestose, analytical standard
C18H32O16
56218
Nystose, analytical standard
C24H42O21