EMAIL THIS PAGE TO A FRIEND
Pharmacognosy magazine
Identity-based High-performance thin Layer Chromatography Fingerprinting Profile and Tumor Inhibitory Potential of Anisochilus carnosus (L.f.) wall Against Ehrlich Ascites Carcinoma.
PMID 26929584
Abstract
Anisochilus carnosus (L.f.) wall belonging to the family Lamiaceae is a plant that is widely used in folk medicine for treating eczema, cold, cough, and fever. In the present study, we explored the anticancer potential of A. carnosus leaves against Ehrlich ascites carcinoma (EAC) and estimated the quantity of luteolin present in various extracts and fractions of A. carnosus by high-performance thin layer chromatography (HPTLC) fingerprinting. Various factors such as tumor volume, tumor cell viability, tumor weight, prolongation of lifespan, and hematological parameters were assessed. We observed a significant lowering in tumor volume, tumor weight, and cell viability in EAC-induced mice following intervention with A. carnosus extracts. Also, there was a considerable prolongation of host lifespan and restoration of hematological parameters to almost normal levels with A. carnosus treatment. HPTLC fingerprinting of various extracts and fractions of A. carnosus along with luteolin as the reference standard revealed the occurrence of luteolin in all tested extracts and fractions of A. carnosus with the highest concentration being reported in the ethanol fraction. A. carnosus exhibits potent anti-tumor potential which can most likely be attributed to the occurrence of different phytochemicals such as phytosterols, terpenoids, and flavonoids in the plant. Further studies to isolate compounds from A. carnosus and understand the mechanism of anti-tumor activity would be worthwhile. EAC induced mice that received A. carnosus treatment exhibited significant reduction in tumor volume, tumor weight and tumor cell viability. Their life span was considerably prolonged. We detected luteolin in A. carnosus aqueous and ethanol extract using HPTLC. Hence, anticancer activity of A. carnosus can be partly attributed to the presence of luteolin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6508
L-Aspartic acid β-hydroxamate, serine racemase inhibitor
C4H8N2O4