EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of gastroenterology. Supplement
Octatropine-methyl-bromide and sulglycotide salt in the short-term treatment of active duodenal ulcer. A double blind endoscopic study of 40 outpatients.
PMID 2694331
Abstract
Forty outpatients with endoscopically confirmed duodenal ulcers were entered in a double blind trial. They were randomly allocated to octatropine-methyl-bromide and sulglycotide salt (GVP) or placebo. The results show that the combination of the two drugs is less efficacious than the two constituent substances taken separately, is not more efficacious than placebo in ulcer healing, and is ineffective with regard to ulcer pain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5181
Anisotropine methyl bromide
C17H32BrNO2