EMAIL THIS PAGE TO A FRIEND
Cytotechnology
Stimulation of fat storage by prostacyclin and selective agonists of prostanoid IP receptor during the maturation phase of cultured adipocytes.
PMID 26946143
Abstract
We have previously shown that cultured adipocytes have the ability to biosynthesize prostaglandin (PG) I
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T2573
Troglitazone, ≥98% (HPLC)
C24H27NO5S