EMAIL THIS PAGE TO A FRIEND
Journal of dental research
Craniofacial and Dental Defects in the Col1a1Jrt/+ Mouse Model of Osteogenesis Imperfecta.
PMID 26951553
Abstract
Certain mutations in the COL1A1 and COL1A2 genes produce clinical symptoms of both osteogenesis imperfecta (OI) and Ehlers-Danlos syndrome (EDS) that include abnormal craniofacial growth, dental malocclusion, and dentinogenesis imperfecta. A mouse model (Col1a1(Jrt)/+) was recently developed that had a skeletal phenotype and other features consistent with moderate-to-severe OI and also with EDS. The craniofacial phenotype of 4- and 20-wk-old Col1a1(Jrt)/+ mice and wild-type littermates was assessed by micro-computed tomography (µCT) and morphometry. Teeth and the periodontal ligament compartment were analyzed by µCT, light microscopy/histomorphometry, and electron microscopy. Over time, at 20 wk, Col1a1(Jrt)/+ mice developed smaller heads, a shortened anterior cranial base, class III occlusion, and a mandibular side shift with shorter morphology in the masticatory region (maxilla and mandible). Col1a1(Jrt)/+ mice also had changes in the periodontal compartment and abnormalities in the dentin matrix and mineralization. These findings validate Col1a1(Jrt)/+ mice as a model for OI and EDS in humans.