EMAIL THIS PAGE TO A FRIEND
Saudi journal of biological sciences
Expression analysis of ClpB/Hsp100 gene in faba bean (Vicia faba L.) plants in response to heat stress.
PMID 26981006
Abstract
Heat stress adversely affects the growth and yield of faba bean crop. Accumulation of ClpB/Hsp100 class of proteins is a critical parameter in induction of acquired heat stress tolerance in plants. Heat-induced expression of ClpB/Hsp100 genes has been noted in diverse plant species. Using primers complementary to soybean ClpB/Hsp100 gene, we analyzed the transcript expression profile of faba bean ClpB/Hsp100 gene in leaves of seedlings and flowering plants and in pollen grains. ClpB/Hsp100 protein accumulation profile was analyzed in leaves of faba bean seedlings using Arabidopsis thaliana cytoplasmic Hsp101 antibodies. The transcript and protein levels of faba bean ClpB/Hsp100 were significantly induced in response to heat stress.