EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Covalent Bonding of Metal-Organic Framework-5/Graphene Oxide Hybrid Composite to Stainless Steel Fiber for Solid-Phase Microextraction of Triazole Fungicides from Fruit and Vegetable Samples.
PMID 26998567
Abstract
A hybrid material of the zinc-based metal-organic framework-5 and graphene oxide (metal-organic framework-5/graphene oxide) was prepared as a novel fiber coating material for solid-phase microextraction (SPME). The SPME fibers were fabricated by covalent bonding via chemical cross-linking between the coating material metal-organic framework-5/graphene oxide and stainless steel wire. The prepared fiber was used for the extraction of five triazole fungicides from fruit and vegetable samples. Gas chromatography coupled with microelectron capture detector (GC-μECD) was used for quantification. The developed method gave a low limit of detection (0.05-1.58 ng g(-1)) and good linearity (0.17-100 ng g(-1)) for the determination of the triazole fungicides in fruit and vegetable samples. The relative standard deviations (RSDs) for five replicate extractions of the triazole fungicides ranged from 3.7 to 8.9%. The method recoveries for spiked fungicides (5, 20, and 50 ng g(-1)) in grape, apple, cucumber, celery cabbage, pear, cabbage, and tomato samples were in the range of 85.6-105.8% with the RSDs ranging from 3.6 to 11.4%, respectively, depending on both the analytes and samples. The metal-organic framework-5/graphene oxide coated fiber was stable enough for 120 extraction cycles without a significant loss of extraction efficiency. The method was suitable for the determination of triazole fungicides in fruit and vegetable samples.