EMAIL THIS PAGE TO A FRIEND
Molecular psychiatry
Monoacylglycerol lipase inhibitors produce pro- or antidepressant responses via hippocampal CA1 GABAergic synapses.
PMID 27001616
Abstract
The probability of suffering the mood disorder depression is up to 30% in women and 15% in men during their life span. Pharmacological options for depression are limited: conventional antidepressants have low efficacy and a delayed onset of action (several weeks). Here we investigate the antidepressant actions of inhibitors of monoacylglycerol lipase (MAGL), the major degradative enzyme of the endocannabinoid 2-arachidonoylglycerol. A low-dose of MAGL inhibitors produces antidepressant effects on acute stress-exposed mice, through glutamatergic synaptic long-term depression (LTD), without significant effects on chronic corticosterone-exposed mice. In contrast, a high-dose of MAGL inhibitors produces pro- or antidepressant effects on acute stress- or chronic corticosterone-exposed mice, respectively, through GABAergic synaptic disinhibition. In the hippocampus, in vivo inhibition of MAGL induces a CB
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML2166
KML29, ≥98% (HPLC)
C24H21F6NO7