EMAIL THIS PAGE TO A FRIEND
Scientific reports
Metformin stimulates IGFBP-2 gene expression through PPARalpha in diabetic states.
PMID 27009398
Abstract
The anti-diabetic drug, metformin, exerts its action through AMP-activated protein kinase (AMPK), and Sirtuin (Sirt1) signaling. Insulin-like growth factor (IGF)-binding protein 2 (IGFBP-2) prevents IGF-1 binding to its receptors, thereby contributing to modulate insulin sensitivity. In this study, we demonstrate that metformin upregulates Igfbp-2 expression through the AMPK-Sirt1-PPARα cascade pathway. In the liver of high fat diet, ob/ob, and db/db mice, Igfbp-2 expression was significantly decreased compared to the expression levels in the wild-type mice (p < 0.05). Upregulation of Igfbp-2 expression by metformin administration was disrupted by gene silencing of Ampk and Sirt1, and this phenomenon was not observed in Pparα-null mice. Notably, activation of IGF-1 receptor (IGF-1R)-dependent signaling by IGF-1 was inhibited by metformin. Finally, when compared to untreated type 2 diabetes patients, the metformin-treated diabetic patients showed increased IGFBP-2 levels with diminished serum IGF-1 levels. Taken together, these findings indicate that IGFBP-2 might be a new target of metformin action in diabetes and the metformin-AMPK-Sirt1-PPARα-IGFBP-2 network may provide a novel pathway that could be applied to ameliorate metabolic syndromes by controlling IGF-1 bioavailability.