EMAIL THIS PAGE TO A FRIEND
Artificial cells, nanomedicine, and biotechnology
Development and characterization of embelin-loaded nanolipid carriers for brain targeting.
PMID 27012597
Abstract
The objective of present work was to enquire the potential use of embelin-loaded nanolipid carriers for brain targeting. The average particle size and polydispersity index (PDI) of optimized formulation (F19) were found to be 152 ± 19.7 nm and 0.143 ± 0.023, respectively. Nanolipid carrier (NLC) was also significantly attenuated pentylenetetrazole (PTZ)-induced biochemical parameters in comparison to plain embelin that results in an increase in the level of malondialdehyde (MDA), nitrite, and reduction in the level of glutathione. From the results, it was concluded that embelin-NLCs developed as a beneficial carrier to achieve sustained release and brain targeting through nasal route.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
464481
2-Octyl-1-dodecanol, 97%
C20H42O