EMAIL THIS PAGE TO A FRIEND
Nihon Heikatsukin Gakkai zasshi
[Effect of HSR-803 on gastrointestinal motility].
PMID 2702276
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0033
Itopride hydrochloride, ≥98% (HPLC)
C20H26N2O4 ·HCl