EMAIL THIS PAGE TO A FRIEND
Cytotechnology
Long term morphological characterization of mesenchymal stromal cells 3D spheroids built with a rapid method based on entry-level equipment.
PMID 27023795
Abstract
Three-dimensional (3D) spheroids of mesenchymal stromal cells (MSC) have been demonstrated to improve a wide range of MSC features, such as multilineage potential, secretion of therapeutic factors, and resistance against hypoxic condition. Accordingly, they represent a promising tool in regenerative medicine for several biological and clinical applications. Many approaches have been proposed to generate MSC spheroids. They usually require specific generation systems, such as rotatory bioreactors or low-attachment plates, and each approach has its own disadvantages. Furthermore, an over-time analysis of morphological homogeneity and architectural stability of the spheroids generated is rarely provided. In this work we adapted the "pellet culture" method to obtain homogenous spheroids of MSC and maintain them in vitro for long term studies. We analysed their outer and inner structure over a 2-month period to provide morphological and architectural information regarding the spheroids generated. Quantitative and qualitative data were obtained using brightfield and confocal microscope imaging coupled to a computational analysis to estimate volume, sphericity, and jagging degree. In addition, histological evaluation was performed to more thoroughly assess the cellular composition and the internal architecture of the 3D spheroids. The results provided show that MSC spheroids generated with the proposed approach are homogeneous and stable, from both morphological and architectural points of view, for a period of at least 15 days, approximately between day 15 and day 30 after their generation. Accordingly, the approach proposed serves as a rapid, cost-effective, and efficient method to generate and maintain MSC spheroids using common entry-level laboratory equipment only.