EMAIL THIS PAGE TO A FRIEND
Journal of the American Veterinary Medical Association
Disappearing food animal drugs.
PMID 2703420
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32173
Ipronidazole, VETRANAL, analytical standard
C7H11N3O2