EMAIL THIS PAGE TO A FRIEND
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Tendon-derived progenitor cells improve healing of collagenase-induced flexor tendinitis.
PMID 27035120
Abstract
Tendinitis is a common and a performance-limiting injury in athletes. This study describes the value of intralesional tendon-derived progenitor cell (TDPC) injections in equine flexor tendinitis. Collagenase-induced tendinitis was created in both front superficial digital flexor (SDF) tendons. Four weeks later, the forelimb tendon lesions were treated with 1 × 10