EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Long noncoding RNA H19 regulates EZH2 expression by interacting with miR-630 and promotes cell invasion in nasopharyngeal carcinoma.
PMID 27040767