EMAIL THIS PAGE TO A FRIEND
The journal of physiological sciences : JPS
Expression of the TRPM6 in mouse placental trophoblasts; potential role in maternal-fetal calcium transport.
PMID 27043350
Abstract
The placenta is required to transport calcium (Ca