EMAIL THIS PAGE TO A FRIEND
The Journal of membrane biology
The Electroporation as a Tool for Studying the Role of Plasma Membrane in the Mechanism of Cytotoxicity of Bisphosphonates and Menadione.
PMID 27044359
Abstract
In this study, the role of the cell plasma membrane as a barrier in the mechanism of the cytotoxicity of nitrogen-containing bisphosphonates and menadione was studied, and the possibility of increasing the efficiency of bisphosphonates and menadione (vitamin K