EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Inhibition of bacterial undecaprenyl pyrophosphate synthase by small fungal molecules.
PMID 27049441
Abstract
Viridicatumtoxin and spirohexaline, small fungal molecules with a tetracyclic scaffold and an additional spirobicyclic ring in common, were found to inhibit bacterial undecaprenyl pyrophosphate (UPP) synthase with IC