EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Fullerene-Free Polymer Solar Cells with over 11% Efficiency and Excellent Thermal Stability.
PMID 27061511
Abstract
A nonfullerene-based polymer solar cell (PSC) that significantly outperforms fullerene-based PSCs with respect to the power-conversion efficiency is demonstrated for the first time. An efficiency of >11%, which is among the top values in the PSC field, and excellent thermal stability is obtained using PBDB-T and ITIC as donor and acceptor, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
900799
ITIC
C94H82N4O2S4
906387
ITIC-Cl
C94H78Cl4N4O2S4