EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Identification and purity determination of benzathine and embonate salts of some beta-lactam antibiotics by thin-layer chromatography.
PMID 2708491