EMAIL THIS PAGE TO A FRIEND
The Journal of urology
Re: Effect of silver oxide/trichloroisocyanuric acid antimicrobial urinary drainage system on catheter-associated bacteriuria.
PMID 2709512
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
176125
Trichloroisocyanuric acid
C3Cl3N3O3
91220
Trichloroisocyanuric acid, technical, ≥95% (CHN)
C3Cl3N3O3