EMAIL THIS PAGE TO A FRIEND
PMID 27123103
Abstract
N-glycosylation is a post-translational protein modification with a wide variety of functions. It has been predicted that R-spondin1 (RSPO1) is N-glycosylated, although this remains unknown. The present study identified that RSPO1 was N-glycosylated at Asn137, and that N-glycosylation of RSPO1 negatively influenced its secretion and enhancing effect on Wnt/β-catenin signaling.