EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Bio-scaffolds produced from irradiated squid pen and crab chitosan with hydroxyapatite/β-tricalcium phosphate for bone-tissue engineering.
PMID 27126171
Abstract
In this study, bio-scaffolds have been developed using irradiated chitosan from different sources - squid pen (RS) and crab shell (RC) - with hydroxyapatite/β-tricalcium phosphate (HA/β-TCP) at a chitosan/HA/β-TCP ratio of 50/30/20. The bio-scaffolds were prepared at two different freezing temperature (-20°C and -80°C) followed by lyophilisation. To enhance the mechanical properties, the bio-scaffolds were cross-linked using sodium tripolyphosphate (TPP) followed by lyophilisation. The composition and morphology of the bio-scaffolds were characterized using XRD, SEM, TEM and μ-CT. The pore size of the porous scaffolds ranged from 90 to 220μm and the scaffolds had 70-80% porosity. The scaffolds had a water uptake ratio of more than 10, and a controlled biodegradation in the range of 30-40%. These results suggest that the physical and biological properties of chitosan-based bio-scaffolds can be a promising biomaterial for bone-tissue regeneration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89050205
SAOS-2 Cell Line human, primary osteogenic sarcoma