EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
CRISPR/Cas9-mediated mutation of PHEX in rabbit recapitulates human X-linked hypophosphatemia (XLH).
PMID 27126636
Abstract
X-linked hypophosphatemia (XLH) is the most common cause of inheritable rickets, with an incidence of 1/20 000 in humans. Inactivation or mutation of the gene PHEX, a phosphate-regulating endopeptidase, leads to hypophosphatemia and defective bone mineralization in XLH patients. Presently, there is no adequate animal model for safety assessments of physiotherapies and drug screening for XLH rickets. In this study, an XLH model was generated via PHEX gene knockout (KO) through coinjection of clustered regularly interspaced short palindromic repeats (CRISPR)-associated protein 9 (Cas9)/sgRNA mRNA into rabbit zygotes. The typical phenotypes of growth retardation, hypophosphatemia, elevated serum FGF23 and bone mineralization were observed in the PHEX KO rabbits but not in normal controls. In summary, for the first time, we have successfully obtained PHEX KO rabbits and recapitulated human XLH using the CRISPR/Cas9 system. This novel XLH rabbit model could be utilized as a drug screening model for XLH prevention and preclinical therapy.