EMAIL THIS PAGE TO A FRIEND
Biochemistry and cell biology = Biochimie et biologie cellulaire
Dimethylmaleic anhydride, a specific reagent for protein amino groups.
PMID 2713129
Abstract
The reagent dimethylmaleic anhydride does not cause a stable modification of thiol compounds under the conditions used for modification of protein amino groups, in contrast to maleic and monomethylmaleic anhydrides, which produce an irreversible modification of sulfhydryl groups. This behavior and the low reactivity toward hydroxyamino acid residues, shown in a previous work, make dimethylmaleic anhydride a specific reagent for protein amino groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D167800
2,3-Dimethylmaleic anhydride, 98%
C6H6O3