EMAIL THIS PAGE TO A FRIEND
Artificial cells, nanomedicine, and biotechnology
In vitro effect of imatinib mesylate loaded on polybutylcyanoacrylate nanoparticles on leukemia cell line K562.
PMID 27133178
Abstract
The study aimed to prepare imatinib mesylate-loaded polybutylcyanoacrylate (PBCA) nanoparticles and evaluate their efficacy on leukemia cell line K562. The formulation was prepared by miniemulsion polymerization technique. Nanoparticles were characterized by dynamic light scattering (DLS), spectrophotometry, Fourier transform infrared spectroscopy (FTIR), dialysis membrane, and 3-(4,5-dimethylthiazol-2-Yl)-2,5-diphenyltetrazolium bromide (MTT) techniques. Nanoscale particles with high encapsulation efficiency (86%) and physical entrapment of drug were observed. In addition, nanoparticles showed suitable drug retention capability and potentiate the cytotoxicity effects of imatinib mesylate. Findings of study suggested PBCA nanoparticles are promising carrier for imatinib mesylate delivery to leukemia cell line K562.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS022173
Imatinib
C29H31N7O