EMAIL THIS PAGE TO A FRIEND
Toxicology
Dispersive liquid-liquid microextraction based on solidification of floating organic drop and high-performance liquid chromatography to the analysis of cocaine's major adulterants in human urine.
PMID 27142991
Abstract
A simple method has been proposed for the determination of cocaine's major adulterants (caffeine, levamisole, lidocaine, phenacetin, diltiazem, and hydroxyzine) in human urine by dispersive liquid-liquid microextraction based on solidification of floating organic drop (DLLME-SFO) in combination with high-performance liquid chromatography - photodiode array detector (HPLC-PDA). The reversed-phase chromatographic separation was obtained with a column C18 extended (250×4.6mm; 5μm; 80Å) in gradient elution mode using acetonitrile-trifluoroacetic acid 0.026% (v,v) (pH=2.5) at 1mLmin
Related Materials