EMAIL THIS PAGE TO A FRIEND
The British journal of dermatology
Dual c-Jun N-terminal kinase-cyclin D1 and extracellular signal-related kinase-c-Jun disjunction in human melanoma.
PMID 27145925
Abstract
Activity of both c-Jun and cyclin D1 is deemed critical for melanoma cell proliferation. This functionality is corroborated by frequently elevated expression and activity of these proteins in human melanomas. Correspondingly, alleviating c-Jun and cyclin D1 function is vital to the success of antimelanoma therapeutics. To understand the role of the c-Jun N-terminal kinase (JNK) signalling pathway in melanoma cell proliferation and survival. The effect of JNK inhibitors SP600125 and JNK-IN-8 on the proliferation and survival of genetically highly representative human melanoma cell lines was studied in assays of proliferation and apoptosis. Changes in c-Jun and cyclin D1 protein and mRNA levels in response to JNK and mitogen-activated protein kinase kinase (MEK) inhibition were investigated through immunoblotting and quantitative reverse-transcription polymerase chain reaction. The effects of JNK and MEK inhibitors on cell-cycle distribution were assessed by flow cytometry. We demonstrate the requirement of JNK signalling in melanoma cell proliferation and survival. While JNK inhibition suppressed the expression and activity of c-Jun, it failed to suppress cyclin D1 levels. Consistently with its inability to downregulate cyclin D1, JNK inhibition failed to induce G1 arrest. In contrast, the blockade of MEK-extracellular signal-regulated kinase (ERK) signalling, although unable to suppress c-Jun activity and expression, paradoxically abated cyclin D1 levels and triggered G1 arrest. This previously unreported dual disconnect between JNK-cyclin D1 and ERK-c-Jun levels was confirmed by concomitant JNK and BRAF inhibition, which suppressed both c-Jun and cyclin D1 levels and exhibited a heightened antiproliferative response. Dual disjunction between JNK-cyclin D1 and ERK-c-Jun signalling forms the basis for further investigation of combined JNK and MAPK signalling blockade as a more effective therapeutic approach in human melanoma.