EMAIL THIS PAGE TO A FRIEND
Chinese journal of integrative medicine
Protective effects of traditional Tibetan medicine Zuo-Mu-A Decoction () on the blood parameters and myocardium of high altitude polycythemia model rats.
PMID 27145942
Abstract
To explore the protective effects of Tibetan medicine Zuo-Mu-A Decoction (, ZMAD) on the blood parameters and myocardium of high altitude polycythemia (HAPC) model rats. Forty male Wistar rats were randomly divided into 4 groups by a random number table, including the normal, model, Rhodiola rosea L. (RRL) and ZMAD groups (10 in each group). Every group was raised in Lhasa to create a HAPC model except the normal group. After modeling, rats in the RRL and the ZMAD groups were administered intragastrically with RRL (20 mL/kg) and ZMAD (7.5 mL/kg) once a day for 2 months, respectively; for the normal and the model groups, 5 mL of distilled water was administered intragastrically instead of decoction. Then routine blood and hematologic rheology parameters were taken, levels of erythropoietin and 8-hydroxy-2'-deoxyguanosine (8-OHdG) were tested, and ultrastructural change in the left ventricular myocardium was observed using transmission electron microscopy. Compared with the model group, ZMAD significantly reduced the red blood cell count, hemoglobin levels, whole blood viscosity at low/middle shear rates, plasma viscosity, erythrocyte electrophoretic time, erythropoietin and 8-OHdG levels, and also increased the erythrocyte deformation index (P<0.05). There was no difference in all results between the RRL and the ZMAD groups. The cardiac muscle fibers were well-protected, mitochondrial matrix swelled mildly and ultrastructure changes were less prominent in the ZMAD group compared with the model group. ZMAD has significant protective effects on the blood parameters against HAPC, and also has the beneficial effect in protecting against myocardial injury.