EMAIL THIS PAGE TO A FRIEND
Histochemistry
Optimization of the histochemical demonstration of DNA using 3-hydroxy-2-naphthoic acid hydrazide and fast blue B.
PMID 2715053
Abstract
Previous methods for the histochemical demonstration of DNA were optimized. p-Toluene sulfonic acid as catalyst for hydrazone formation between the aldehydes generated after Feulgen hydrolysis and 3-hydroxy-2-naphthoic acid hydrazide (NAH) was used instead of acetic acid. Modifications of the conditions of the coupling reaction with Fast Blue B reduced the background staining. The optimized histochemical staining method for DNA (NAH-FB-DNA staining) can be performed easily and reproducibly. Without prior Feulgen hydrolysis the optimized method can also be used for the histochemical demonstration of reactive carbonyls undissolved under the given histochemical conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
55940
3-Hydroxy-2-naphthoic hydrazide, ≥98.0%
C11H10N2O2
D9805
Fast Blue B Salt, Dye content ~95 %
C14H12Cl4N4O2Zn