EMAIL THIS PAGE TO A FRIEND
Drug and alcohol dependence
Nationwide increase in hospitalizations for heroin-related soft tissue infections: Associations with structural market conditions.
PMID 27155756
Abstract
Little is known about trends in national rates of injection-related skin and soft tissue infections (SSTI) and their relationship to the structural risk environment for heroin users. Use of Mexican-sourced "Black Tar" heroin, predominant in western US states, may have greater risk for SSTI compared with eastern US powder heroin (Colombian-sourced) due to its association with non-intravenous injection or from possible contamination. Using nationally representative hospital admissions data from the Nationwide Inpatient Sample and heroin price and purity data from the Drug Enforcement Administration, we looked at rates of hospital admissions for opiate-related SSTI (O-SSTI) between 1993 and 2010. Regression analyses examined associations between O-SSTI and heroin source, form and price. Hospitalization rates of O-SSTI doubled from 4 to 9 per 100,000 nationally between 1993 and 2010; the increase concentrated among individuals aged 20-40. Heroin market features were strongly associated with changes in the rate of SSTI. Each $100 increase in yearly heroin price-per-gram-pure was associated with a 3% decrease in the rate of heroin-related SSTI admissions. Mexican-sourced-heroin-dominant cities had twice the rate of O-SSTI compared to Colombian-sourced-heroin-dominant cities. Heroin-related SSTI are increasing and structural factors, including heroin price and source-form, are associated with higher rates of SSTI hospital admissions. Clinical and harm reduction efforts should educate heroin users on local risk factors, e.g., heroin type, promote vein health strategies and provide culturally sensitive treatment services for persons suffering with SSTI.