EMAIL THIS PAGE TO A FRIEND
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Inflammatory Osteoclasts Prime TNFα-Producing CD4
PMID 27161765
Abstract
Bone destruction is a hallmark of chronic rheumatic diseases. Although the role of osteoclasts in bone loss is clearly established, their implication in the inflammatory response has not been investigated despite their monocytic origin. Moreover, specific markers are lacking to characterize osteoclasts generated in inflammatory conditions. Here, we have explored the phenotype of inflammatory osteoclasts and their effect on CD4