EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
Cancer Stem-like Properties in Colorectal Cancer Cells with Low Proteasome Activity.
PMID 27166395
Abstract
One of the main reasons for cancer treatment resistance is the existence of cancer stem-like cells (CSCs). Here, we elucidated the relationship between low proteasome activity cells (LPACs) and CSCs. The human colorectal cancer cell lines HCT116, SW480, DLD1, and KM12SM were engineered to stably express a green fluorescent molecule fused to the degron of ornithine decarboxylase, resulting in an accumulation of the fluorescence in LPACs. LPACs were isolated by flow cytometry. Treatment resistance (radio- and chemotherapy) and the capacity of LPACs to act as CSCs were analyzed. Microarray analysis was performed to reveal genes related to treatment resistance. The prognostic impact of potent genes was examined in 190 patients with colorectal cancer. LPACs had a significantly increased capacity for radioresistance and chemoresistance (5-fluorouracil and oxaliplatin), significantly lower reactive oxygen species activity, and significantly increased sphere formation capacity compared with non-LPACs. The number of cells in the G LPACs have characteristic treatment resistance and act as CSCs in colorectal cancer. In addition, EID3 is one of the potential regulators of treatment resistance in colorectal cancer and may be a potential therapeutic target. Clin Cancer Res; 22(21); 5277-86. ©2016 AACR.