EMAIL THIS PAGE TO A FRIEND
Clinical oral investigations
The influence of geranylgeraniol on microvessel sprouting after bisphosphonate substitution in an in vitro 3D-angiogenesis assay.
PMID 27170294
Abstract
Recent studies focused on angiogenesis in the pathophysiology of bisphosphonate-associated osteonecrosis of the jaws (BP-ONJ) and identified geranylgeraniol (GGOH) as a feasible option for BP-ONJ therapy. This study investigated the influence of GGOH on microvessel sprouting after BP-incubation in vitro. Ten experimental set-ups were randomly designed in an in vitro 3D-angiogenesis assay. Two groups included HUVEC cell spheroids with and without (±) GGOH substitution as controls and eight groups pairwise contained either clodronate or the nitrogen-containing bisphosphonates (N-BP) ibandronate, pamidronate, and zoledronate ± GGOH. The size of the cell spheroids including the outbranching sprouts (SpS) as well as the density (SpD) and length of the sprouts (SpL) were analyzed by a grid system after 0, 24, 48, and 72 h. For controls and NN-BP clodronate, no significant differences at any tested parameter and any point of measurement could be detected within the experimental set-ups ± GGOH (p each ≥0.05). For N-BP ibandronate, the experimental set-ups +GGOH showed a significantly increased SpS, SpD, and SpL after 48 and 72 h (p each ≤0.002) compared to the experimental set-ups -GGOH. For N-BPs pamidronate and zoledronate, the experimental set-ups + GGOH demonstrated a significantly increased SpS, SpD, and SpL after 24, 48, and 72 h (p each ≤0.001) compared to the experimental set-ups -GGOH. The strong negative influence of N-BPs on microvessel sprouting could be significantly reversed by GGOH. Since supportive therapeutic options for BP-ONJ are lacking, GGOH might be a promising substitute for BP-ONJ prevention and therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3278
Geranylgeraniol, ≥85% (GC)
C20H34O