EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
Targeted inactivation of murine Ddx3x: essential roles of Ddx3x in placentation and embryogenesis.
PMID 27179789
Abstract
The X-linked DEAD-box RNA helicase DDX3 (DDX3X) is a multifunctional protein that has been implicated in gene regulation, cell cycle control, apoptosis, and tumorigenesis. However, the precise physiological function of Ddx3x during development remains unknown. Here, we show that loss of Ddx3x results in an early post-implantation lethality in male mice. The size of the epiblast marked by Oct3/4 is dramatically reduced in embryonic day 6.5 (E6.5) Ddx3x