EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Gastroenterologie
European Association for Gastroenterology and Endoscopy, 21st congress. Graz, Austria, May 3-6, 1989. Abstracts.
PMID 2718537
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV45827
Anti-CHAF1B antibody produced in rabbit, IgG fraction of antiserum