EMAIL THIS PAGE TO A FRIEND
Journal of experimental pharmacology
Effects of an oral dose of l-glutamic acid on circulating neurotransmitters: Possible roles of the C1(Ad) and the A5(NA) pontomedullary nuclei.
PMID 27186090
Abstract
Investigation of the effects of an oral administration of a small dose of l-glutamic acid on the two peripheral sympathetic branches (neural and adrenal) of the autonomic nervous system. Circulating neurotransmitters and cardiovascular parameters were assessed in 28 healthy volunteers before and after the administration of 500 mg of l-glutamic acid or placebo. The drug triggered a significant and sustained enhancement of the noradrenaline and dopamine circulating levels which were paralleled and positively correlated with the diastolic blood pressure increases. Conversely, both platelet and plasma serotonin showed significant falls throughout the test. Significant positive correlations were registered between noradrenaline, dopamine, and noradrenaline/dopamine ratio versus diastolic blood pressure but not versus systolic blood pressure or heart rate. The above results allowed us to postulate that the drug provoked a significant enhancement of peripheral neural sympathetic activity and the reduction of adrenal sympathetic and parasympathetic drives. Both sympathetic branches are positively correlated with the A5 noradrenergic and the C1 adrenergic pontomedullary nuclei, which interchange inhibitory axons that act at post-synaptic α2 inhibitory autoreceptors. In addition, we discussed the mechanisms able to explain why the drug acted preferentially at the A5 noradrenergic rather than the C1 adrenergic nuclei.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442291
Octadecylsilane, 97%
C18H40Si