EMAIL THIS PAGE TO A FRIEND
Journal of exposure science & environmental epidemiology
Is daily exposure to ozone associated with respiratory morbidity and lung function in a representative sample of schoolchildren? Results from a panel study in Greece.
PMID 27189255
Abstract
Previous time series or panel studies of asthmatics have reported respiratory health effects following short-term exposure to ozone (O