EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Local and skin resorptive action of N,N-diethylchloracetamide].
PMID 2722018
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
250996
2-Chloro-N,N-diethylacetamide, 97%
C6H12ClNO