EMAIL THIS PAGE TO A FRIEND
The Cleft palate-craniofacial journal : official publication of the American Cleft Palate-Craniofacial Association
Early Craniofacial Defects in Zebrafish That Have Reduced Function of a Wnt-Interacting Extracellular Matrix Protein, Tinagl1.
PMID 27243669
Abstract
Tinagl1 has a weak genetic association with craniosynostosis, but its functions in cartilage and bone development are unknown. Knockdown of Tinagl1 in zebrafish embryos allowed an initial characterization of its potential effects on craniofacial cartilage development and a test of whether these effects could involve Wnt signaling. Tinagl1 knockdown resulted in dose-dependent reductions and defects in ventral pharyngeal arch cartilages as well as the ethmoid plate, a zebrafish correlate to the palate. These defects could be correlated to reduced numbers of cranial neural crest cells in the pharyngeal arches and could be reproduced with comanipulation of Tinagl1 and Wnt3a by morpholino-based knockdown. These results suggest that Tinagl1 is required early in the proliferation or migration of cranial neural crest cells and that its effects are mediated via Wnt3a signaling. Because Wnt3a is among the Wnts that contribute to nonsyndromic cleft lip and cleft palate in mouse and man, further investigation of Tinagl1 may help to elucidate mechanisms underlying these disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323616
3′-Deoxythymidine, 98%
C10H14N2O4