EMAIL THIS PAGE TO A FRIEND
eLife
Phylogenetic sleuthing reveals pair of paralogous genes.
PMID 27244557
Abstract
The complex evolutionary history of the IFIT family of antiviral genes has been shaped by continuous interactions between mammalian hosts and their many viruses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10466 CompoZr® Knockout ZFN Kit, Human IFIT1B (NM_001010987)