EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
Study of the Vapor Phase Over Fusarium Fungi Cultured on Various Substrates.
PMID 27253722
Abstract
The compositions of volatile organic compounds (VOCs) emitted by Fusarium fungi (F. langsethiae, F. sibiricum, F. poae, and F. sporotrichioides) grown on two nutritive substrates: potato sucrose agar (PSA) and autoclaved wheat kernels (WK) were investigated. The culturing of fungi and study of their VOC emissions were performed in chromatographic vials at room temperature (23 - 24 °C) and the VOCs were sampled by a solid-phase microextraction on a 85 μm carboxen/polydimethylsiloxane fiber. GC/MS was performed using a 60-m HP-5 capillary column. Components of the VOC mixture were identified by electron impact mass spectra and chromatographic retention indices (RIs). The most abundant components of the VOC mixture emitted by Fusarium fungi are EtOH, AcOH, (i) BuOH, 3-methylbutan-1-ol, 2-methylbutan-1-ol, ethyl 3-methylbutanoate, terpenes with M 136, sesquiterpenes with M 204 (a total of about 25), and trichodiene. It was found that the strains grown on PSA emit a wider spectrum and larger amount of VOCs compared with those grown on wheat kernels. F. langsethiae strain is the most active VOC producer on both substrates. The use of SPME and GC/MS also offers the potential for differentiation of fungal species and strains.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
24241-U Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
25461 Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
24246 Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
25467 Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
54041-U Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
29753-U Carboxen® 569, O.D. × L 1/4 in. × 3 1/2, glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29534-U Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
54293-U Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: (300 mg), cartridge volume 3 mL, pkg of 10 ea
54294-U Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: 300 mg, cartridge size 3 mL, pkg of 50 ea
10263 Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
10264 Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
11324-U Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
10269 Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
11048-U Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
11072-U Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
10477-U Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
10478-U Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
10471 Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
23446-U ORBO-93 Sorbent Tube, matrix Carboxen® 1000, ORBO tube I.D. × L 6 mm × 95 mm, Bed B 90 mg, Bed A 180 mg, 60/80 mesh, pkg of 25 ea
20355 ORBO 78 HBR on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
28718-U Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea